Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Laemmli SDS Sample Buffer, with PyroninY, 4X, 50 mL
Supplier: Thermo Fisher Scientific
Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Catalog # J62115.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Pink
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Specifications
Specifications
- Pk:50 mL
- Description:Laemmli SDS Sample Buffer, with PyroninY, 4X
- Cat. No.:J62115.AD
- Physical form:Liquid
- Appearance:Pink



