Order Entry
Export
Orders LinkContactUsLinkComponent
Acrylamide 30% in aqueous solution

Acrylamide 30% in aqueous solution

Glass bottle, 1 L
Catalog # J62100.K2
Health
Irritant
Acrylamide 30% in aqueous solution
Catalog # J62100.K2
CAS Number:  79-06-1
Acrylamide 30% in aqueous solution
Acrylamide 30% in aqueous solution

Glass bottle, 1 L

CAS Number:  79-06-1
Price
241,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Merck Index:14,129
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:3426
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications