Acrylamide 30% in aqueous solution
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Acrylamide 30% in aqueous solution
Catalog # J62100.AP
Supplier: Thermo Fisher Scientific
CAS Number: 79-06-1
Jump to:
Product Details & Documents
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Merck Index:14,129
- Storage Temperature:Refrigerator
- EINECS:201-173-7
- UN:3426
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



