Order Entry
Export
Orders LinkContactUsLinkComponent
 

Neutral red ACS

Glass bottle for solids, 50 g
Catalog # J62643.18
Neutral red ACS
Catalog # J62643.18
CAS Number:  553-24-2
Neutral red ACS
Neutral red ACS

Glass bottle for solids, 50 g

CAS Number:  553-24-2
Price
45,00 €
Log in for availability
1 * 50 g
Product Details & Documents

CAS No.: 553-24-2 Molecular Formula: C15H17ClN4 Formula Weight: 288.78 Physical Form: Powder Appearance: Dark green to black Melting Point: 290° dec. Beilstein Reference: 3918740 Merck Reference: 14,6488 MDL No.: MFCD00012651

Nuclear counterstain useful in pH range 6.8 - 8.0

Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • MDL Number:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:14,6488
  • Melting Pt:290 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:209-035-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications