Order Entry
Export
Orders LinkContactUsLinkComponent
Auramine O ≥80%

Auramine O ≥80%

Glass bottle for solids, 50 g
Catalog # J60343.18
Toxic
Health
Auramine O ≥80%
Catalog # J60343.18
CAS Number:  2465-27-2
Auramine O ≥80%
Auramine O ≥80%

Glass bottle for solids, 50 g

CAS Number:  2465-27-2
Price
75,50 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow
  • CAS Number:2465-27-2
  • Formula:C₁₇H₂₂ClN₃
  • MDL Number:MFCD00012484
  • Molecular Weight:303.84 g/mol
  • Melting Pt:250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:219-567-2
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications