Order Entry
Export
Orders LinkContactUsLinkComponent
Acrylamide 40% in aqueous solution

Acrylamide 40% in aqueous solution

Glass bottle, 500 mL
Catalog # J62480.AP
Health
Irritant
Acrylamide 40% in aqueous solution
Catalog # J62480.AP
CAS Number:  79-06-1
Acrylamide 40% in aqueous solution
Acrylamide 40% in aqueous solution

Glass bottle, 500 mL

CAS Number:  79-06-1
Price
125,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Merck Index:14,129
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:3426
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications