Order Entry
Export
Orders LinkContactUsLinkComponent
TRIS-glycine native sample buffer (4X), pH 8,6

TRIS-glycine native sample buffer (4X), pH 8,6

TRIS-Glycine Native Sample Buffer (4X), 8,6, 25 mL
Catalog # J61864.AC
TRIS-glycine native sample buffer (4X), pH 8,6
Catalog # J61864.AC
CAS Number:  
TRIS-glycine native sample buffer (4X), pH 8,6
TRIS-glycine native sample buffer (4X), pH 8,6

TRIS-Glycine Native Sample Buffer (4X), 8,6, 25 mL

CAS Number:  
Price
71,30 €
Log in for availability
1 * 25 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless to yellow
MDL No.: MFCD00236360
Applications: For preparation of protein samples for native gel electrophoresis

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    TRIS-Glycine Native Sample Buffer (4X)
  • pH value:
    8,6
  • Cat. No.:
    J61864.AC
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236360
  • Appearance:
    Colourless to yellow

Specifications