Order Entry
Export
Orders LinkContactUsLinkComponent
 

Histamine diphosphate 98%

25 g
  J61088.14
 
 
 
Histamine diphosphate 98%
  J61088.14
 :  51-74-1
Histamine diphosphate 98%
Histamine diphosphate 98%

25 g

 :  51-74-1
 
288,00 €
   
1 * 25 g
 

Purity: 99+% CAS No.: 51-74-1 Molecular Formula: C5H9N3.2H3PO4 Formula Weight: 307.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 128-132° MDL No.: MFCD00064316

Applications: Endogenous H1 and H2 agonist

 :  2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate
  •  :51-74-1
  •  :C₅H₉N₃·2H₃PO₄
  •  :MFCD00064316
  •  :307.14 g/mol
  •  :14,4719
  •  :128…132 °C
  •  :Refrigerator
  •  :200-118-4
 

 

  • Pk:
    25 g