Order Entry
Export
Orders LinkContactUsLinkComponent
 

Histamine diphosphate 98%

5 g
Catalog # J61088.06
Health
Corrosive
Irritant
Histamine diphosphate 98%
Catalog # J61088.06
CAS Number:  51-74-1
Histamine diphosphate 98%
Histamine diphosphate 98%

5 g

CAS Number:  51-74-1
Price
87,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99+% CAS No.: 51-74-1 Molecular Formula: C5H9N3.2H3PO4 Formula Weight: 307.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 128-132° MDL No.: MFCD00064316

Applications: Endogenous H1 and H2 agonist

Synonyms:  2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate
  • CAS Number:51-74-1
  • Formula:C₅H₉N₃·2H₃PO₄
  • MDL Number:MFCD00064316
  • Molecular Weight:307.14 g/mol
  • Merck Index:14,4719
  • Melting Pt:128…132 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-118-4
Specifications

Specifications

  • Pk:
    5 g

Specifications