Order Entry
Export
Orders LinkContactUsLinkComponent
Colchicine ≥98%

Colchicine ≥98%

Glass bottle for solids, 1 g
Catalog # J61072.03
Colchicine ≥98%
Catalog # J61072.03
CAS Number:  64-86-8
Colchicine ≥98%
Colchicine ≥98%

Glass bottle for solids, 1 g

CAS Number:  64-86-8
Price
139,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:14,2471
  • Melting Pt:∼ 150 °C (dec.)
  • Density:1.32 g/cm³ (20 °C)
  • UN:1544
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications