Order Entry
Export
Orders LinkContactUsLinkComponent
TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3

TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3

TRIS-Tricine-SDS running buffer (10X), cathode buffer, 8.3, 1 L
Catalog # J60992.K2
TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3
Catalog # J60992.K2
CAS Number:  
TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3
TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3

TRIS-Tricine-SDS running buffer (10X), cathode buffer, 8.3, 1 L

CAS Number:  
Price
201,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Physical Form: Liquid
Applications: For TRIS-Tricine SDS gel electrophoresis

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    TRIS-Tricine-SDS running buffer (10X), cathode buffer
  • pH value:
    8.3
  • Cat. No.:
    J60992.K2
  • Physical form:
    Liquid

Specifications