TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3
TRIS-Tricine-SDS running buffer (10X), cathode buffer, 8.3, 500 mL
Supplier: Thermo Fisher Scientific
TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3
Catalog # J60992.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Applications: For TRIS-Tricine SDS gel electrophoresis
Documents
Specifications
Specifications
- Pk:500 mL
- Description:TRIS-Tricine-SDS running buffer (10X), cathode buffer
- pH value:8.3
- Cat. No.:J60992.AP
- Physical form:Liquid



