Order Entry
Export
Orders LinkContactUsLinkComponent
Alkaline phosphatase buffer-1 (5X)

Alkaline phosphatase buffer-1 (5X)

Alkaline phosphatase buffer (5X), 250 mL
Catalog # J62907.AK
Alkaline phosphatase buffer-1 (5X)
Catalog # J62907.AK
CAS Number:  
Alkaline phosphatase buffer-1 (5X)
Alkaline phosphatase buffer-1 (5X)

Alkaline phosphatase buffer (5X), 250 mL

CAS Number:  
Price
86,20 €
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Clear liquid

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Alkaline phosphatase buffer (5X)
  • Cat. No.:
    J62907.AK
  • Physical form:
    Liquid
  • Appearance:
    Clear liquid

Specifications