Dextran, M.W. ~ 250,000
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White Merck Reference: 14,2948 MDL No.: MFCD00130935
- CAS Number:9004-54-0
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00130935
- Merck Index:14,2948
- Storage Temperature:Ambient
- EINECS:232-677-5
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



