RBC lysis buffer for human
RBC lysis buffer for human, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Applications: For lysis of human red blood cells.
Documents
Specifications
Specifications
- Pk:500 mL
- Description:RBC lysis buffer for human
- Cat. No.:J62990.AP
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- Appearance:Colourless



