Order Entry
Export
Orders LinkContactUsLinkComponent
 

RBC lysis buffer for human

RBC lysis buffer for human, 250 mL
Catalog # J62990.AK
RBC lysis buffer for human
Catalog # J62990.AK
CAS Number:  
RBC lysis buffer for human
RBC lysis buffer for human

RBC lysis buffer for human, 250 mL

CAS Number:  
Price
75,30 €
Log in for availability
1 * 250 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Applications: For lysis of human red blood cells.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    RBC lysis buffer for human
  • Cat. No.:
    J62990.AK
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications