Order Entry
Export
Orders LinkContactUsLinkComponent
Phosphate-citrate Buffer (5X)

Phosphate-citrate Buffer (5X)

Phosphate-citrate buffer (5X), 2,5, 250 ml
Catalog # J62982.AK
Phosphate-citrate Buffer (5X)
Catalog # J62982.AK
Supplier:  
CAS Number:  
Phosphate-citrate Buffer (5X)
Phosphate-citrate Buffer (5X)

Phosphate-citrate buffer (5X), 2,5, 250 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Phosphate-Citrate Buffer (5X) 1 * 100 mL Catalog #J62982.AE

Product J62982.AE is a replacement for J62982.AK but varies by Brand, Packaging or Quantity.

Product Details & Documents

Phosphate-Citrate Buffer (5X) is used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.

Contains 1M sodium phosphate and 0.5M sodium citrate.

Specifications

Specifications

  • Pk:
    250 ml
  • Description:
    Phosphate-citrate buffer (5X)
  • pH value:
    2,5
  • Cat. No.:
    J62982.AK
  • MDL No.:
    MFCD00131859

Specifications