Order Entry
Export
Orders LinkContactUsLinkComponent
Phosphate-citrate Buffer (5X)

Phosphate-citrate Buffer (5X)

Phosphate-citrate buffer (5X), 2,5, 100 mL
Catalog # J62982.AE
Phosphate-citrate Buffer (5X)
Catalog # J62982.AE
CAS Number:  
Phosphate-citrate Buffer (5X)
Phosphate-citrate Buffer (5X)

Phosphate-citrate buffer (5X), 2,5, 100 mL

CAS Number:  
Price
74,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Phosphate-Citrate Buffer (5X) is used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.

Contains 1M sodium phosphate and 0.5M sodium citrate.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Phosphate-citrate buffer (5X)
  • pH value:
    2,5
  • Cat. No.:
    J62982.AE
  • MDL No.:
    MFCD00131859

Specifications