TE buffer, pH 7,6, RNAse free
TE buffer, RNAse free, 7,6, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. TE buffer functions to solubilise DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.
Physical Form: Liquid
MDL No.: MFCD00236359
Specifications
Specifications
- Pk:250 mL
- Description:TE buffer, RNAse free
- pH value:7,6
- Cat. No.:J62285.AK
- Storage temperature:Ambient temperature
- Physical form:Liquid
- MDL No.:MFCD00236359



