Order Entry
Export
Orders LinkContactUsLinkComponent
TE buffer, pH 7,6, RNAse free

TE buffer, pH 7,6, RNAse free

TE buffer, RNAse free, 7,6, 125 mL
Catalog # J62285.XE
TE buffer, pH 7,6, RNAse free
Catalog # J62285.XE
CAS Number:  
TE buffer, pH 7,6, RNAse free
TE buffer, pH 7,6, RNAse free

TE buffer, RNAse free, 7,6, 125 mL

CAS Number:  
Price
67,20 €
Log in for availability
1 * 125 mL
Product Details & Documents

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. TE buffer functions to solubilise DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

Physical Form: Liquid
MDL No.: MFCD00236359

Specifications

Specifications

  • Pk:
    125 mL
  • Description:
    TE buffer, RNAse free
  • pH value:
    7,6
  • Cat. No.:
    J62285.XE
  • Storage temperature:
    Ambient temperature
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236359

Specifications