Order Entry
Export
Orders LinkContactUsLinkComponent
 

Buffers, MOPS, 1,0 M solutions

MOPS buffer solution pH 5,5 (1 mol/l), 5,5, Glass bottle, 500 mL
Catalog # J62261.AP
Buffers, MOPS, 1,0 M solutions
Catalog # J62261.AP
CAS Number:  
Buffers, MOPS, 1,0 M solutions
Buffers, MOPS, 1,0 M solutions

MOPS buffer solution pH 5,5 (1 mol/l), 5,5, Glass bottle, 500 mL

CAS Number:  
Price
134,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    MOPS buffer solution pH 5,5 (1 mol/l)
  • pH value:
    5,5
  • Cat. No.:
    J62261.AP
  • Odour/scent:
    Odourless
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006183
  • Appearance:
    Colourless
  • CAS No.:
    1132-61-2

Specifications