Buffers, MOPS, 1,0 M solutions
MOPS buffer solution pH 5,5 (1 mol/l), 5,5, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Description:MOPS buffer solution pH 5,5 (1 mol/l)
- pH value:5,5
- Cat. No.:J62261.AK
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00006183
- Appearance:Colourless
- CAS No.:1132-61-2



