Accessories for Loopster basic and digital
Tube attachment for 12×30 ml tubes with Ø25 mm
Catalog # 444-0853
Supplier: IKA
Jump to:
Product Details & Documents
Specifications
- Description:Tube attachment for 12×30 ml tubes with Ø25 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø25 mm for use with Universal discSpare clips with Ø25 mm for use with Universal disc444-0952Log in for availability1 * 12 items - 48,00 €
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø12 mm for use with Universal discSpare clips with Ø12 mm for use with Universal disc444-0857Log in for availability1 * 12 items - 48,00 €
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø10 mm for use with Universal discSpare clips with Ø10 mm for use with Universal disc444-0856Log in for availability1 * 12 items - 48,00 €
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø28 mm for use with Universal discSpare clips with Ø28 mm for use with Universal disc444-0953Log in for availability1 * 12 items - 48,00 €
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø16 mm for use with Universal discSpare clips with Ø16 mm for use with Universal disc444-0951Log in for availability1 * 12 items - 48,00 €



