System PAGE/Blotting Dual Midi-Vertical CE 2-place gel wrap casting bundle gel dimensions 15x10cm. Includes: lower buffer reservoir stirring coral, 1 dual capacity core, safety cover, power leads, 2 blotting cassettes, sponge pads, 2 glass plate se 5 1 * 1 items
Catalog # CBSCDCX800HC1520
System PAGE/Blotting Dual Midi-Vertical CE 2-place gel wrap casting bundle gel dimensions 15x10cm. Includes: lower buffer reservoir stirring coral, 1 dual capacity core, safety cover, power leads, 2 blotting cassettes, sponge pads, 2 glass plate se 5 1 * 1 items
Catalog # CBSCDCX800HC1520