ContactUsLinkComponent
System PAGE/Blotting Dual Midi-Vertical 4-place gel casting chamber bundle gel dimensions 15x10cm. Includes: lower buffer reservoir stirring coral, 1 dual capacity core, safety cover, power leads, 2 blotting cassettes, sponge pads, 4 glass plate se 7 1 * 1 items
Catalog # CBSCDCX80MHC0712
Supplier:  Expedeon
System PAGE/Blotting Dual Midi-Vertical 4-place gel casting chamber bundle gel dimensions 15x10cm. Includes: lower buffer reservoir stirring coral, 1 dual capacity core, safety cover, power leads, 2 blotting cassettes, sponge pads, 4 glass plate se 7 1 * 1 items
Catalog # CBSCDCX80MHC0712
Supplier:  Expedeon