Orders LinkContactUsLinkComponent
Stirrer blades
Stirrer blades
Anchor stirrer blades, PTFE, with stiffened metal core, for SVL reaction vessels, 54 mm
Catalog # BIBB588-98
Supplier:   SciLabware
Stirrer blades
Catalog # BIBB588-98
Supplier:  SciLabware
Stirrer blades
Stirrer blades
Anchor stirrer blades, PTFE, with stiffened metal core, for SVL reaction vessels, 54 mm
Price
366,00 €
Log in for availability
1 * 5 items
Product Details & Documents
A range of PTFE coated stirrer heads designed to screw into the threads on SVL® glass stirrer shafts.
  • Head simply screws into the end of the shaft
  • Anchor and helix types have a PTFE coated metal core
Caution: Stirrers must be rotated in a clockwise direction to prevent unscrewing of the thread.
Specifications
  • Type:
    Anchor stirrer blades, PTFE, with stiffened metal core, for SVL reaction vessels
  • Width:
    54 mm
  • Seq:
    012