Order Entry
Export
Orders LinkContactUsLinkComponent
Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP

Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP

2 ml, 0,006 ± ml, 330 mm, Orange
Catalog # 612-4786
Supplier:   Brand
Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP
Catalog # 612-4786
Supplier:  Brand
Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP
Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP

2 ml, 0,006 ± ml, 330 mm, Orange

Price
91,80 €
Log in for availability
1 * 12 items
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
  • Incl. USP batch certificate, also available with USP individual certificate on request

Certifications: DIN EN ISO 648; DE-M marking.

Specifications
  • Colour code:
    Orange
  • Capacity:
    2 ml
  • Length:
    330 mm
  • Tolerance:
    0,006 ± ml