0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
GHS HAZARDOUS MATERIALS SIGNS No. 3 13x 1 * 10 items
LBIN9910-GHS03
:
LAB IN
GHS HAZARDOUS MATERIALS SIGNS No. 3 13x 1 * 10 items
LBIN9910-GHS03
:
LAB IN