Orders LinkContactUsLinkComponent
Objectives for stereo zoom microscopes
Objective, 128 mm
Catalog # 630-2042
Supplier:   MOTIC
Objectives for stereo zoom microscopes
Catalog # 630-2042
Supplier:  MOTIC
Objectives for stereo zoom microscopes
Objectives for stereo zoom microscopes
Objective, 128 mm
Price
109,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    0,75×
  • Description:
    Objective
  • Seq:
    102
  • Working distance:
    128 mm