Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Microscope Objectives
Objectives for stereo zoom microscopes
Objectives for stereo zoom microscopes
Print
Objectives for stereo zoom microscopes
Objective, 56,3 mm
Catalog # 630-2040
Supplier:
MOTIC
Objectives for stereo zoom microscopes
Catalog # 630-2040
Supplier:
MOTIC
Expand image
Return to Top
Objectives for stereo zoom microscopes
Objective, 56,3 mm
Specifications
Add to cart
Jump to:
Specifications
Price
109,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Magnification:
1,5×
Description:
Objective
Seq:
103
Working distance:
56,3 mm