Orders LinkContactUsLinkComponent
Disposable cannulas
Disposable cannulas
Disposable cannulas, G23, Blue, 80 mm, 0,6×80 mm
Catalog # EHRS15884480
Supplier:   EHRHARDT SOEHNE
Disposable cannulas
Catalog # EHRS15884480
Supplier:  EHRHARDT SOEHNE
Disposable cannulas
Disposable cannulas
Disposable cannulas, G23, Blue, 80 mm, 0,6×80 mm
Price
55,40 €
Log in for availability
1 * 100 items
Product Details & Documents
Disposable cannulas.
  • Pyrogen-free, not toxic
  • With lancet cut
  • EO-sterilisation
Documents
Specifications
  • Ø×L:
    0,6×80 mm
  • Gauge:
    G23
  • Colour:
    Blue
  • Description:
    Disposable cannulas
  • Length:
    80 mm
  • Seq:
    13