Order Entry
Export
Orders LinkContactUsLinkComponent
 

Hollow fiber filter modules, MicroKros®

Box of 1 module, PS membrane, wet packaging, pore rating MWCO 500 kD, surface area 59 cm²
Catalog # SPECC04-S500-05-P
Hollow fiber filter modules, MicroKros®
Catalog # SPECC04-S500-05-P
Hollow fiber filter modules, MicroKros®

Box of 1 module, PS membrane, wet packaging, pore rating MWCO 500 kD, surface area 59 cm²

Price
295,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Designed for gentle tangential flow filtration of small volumes ranging from 1 to 100 ml. MicroKros® modules are typically operated by a peristaltic pump, these devices also offer the unique advantage of manual operation for quick and convenient separations for small volumes.

  • 5 to 30 minute separations
  • Two modes: Pump operation and Manual operation
  • Gentle process for increased viability
  • 90 to 99% product purity and 85 to 95% product recovery

 : The syringe accessory kit includes 10 × 5 ml syringes, 15 × male Luer cap, 10 × male Luer to 3/16" hose barb fitting, 5 × check valve, 10 × female Luer T connector.

Specifications
  • Description:
    Box of 1 module, PS membrane, wet packaging, pore rating MWCO 500 kD, surface area 59 cm²
  • Seq:
    75
  • Cat. No.:
    SPECC04-S500-05-P
  • Inlet/outlet connections:
    Female Luer lock/male Luer lock
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • MicroKros® syringe accessory kit
    MicroKros® syringe accessory kit
    SPECACPX-400-01N
    Log in for availability
    1 * 1 KIT - 413,00 €