Bottles, narrow neck, with thread, SYSTEM PLUS® 32
315 ml, DIN 32
215-3259
Pure aluminium.
- Unbreakable, odorless
- Very good temperature resistance
- Leakproof with tamper-evident screw caps (must be odered separately)
- UN-approved
Suitable for storage and transport of highly aggessive liquids.
- Thread:DIN 32
- Capacity:315 ml