ContactUsLinkComponent
VERSA 110 NAP/PCR Set-up Workstation including: VD110 - Deck: 8 position deck with tip priming, waste dispensing and tip dispenser AS100 - VERSA110 - Features one robotic arm for liquid handling and deck LD2M-1CDSPTC -Dual Module Manifold - Single Ch 1 * 1 ST
Catalog # IBSAAU-VER-110-NAP
Supplier:  IBS Stock Items
VERSA 110 NAP/PCR Set-up Workstation including: VD110 - Deck: 8 position deck with tip priming, waste dispensing and tip dispenser AS100 - VERSA110 - Features one robotic arm for liquid handling and deck LD2M-1CDSPTC -Dual Module Manifold - Single Ch 1 * 1 ST
Catalog # IBSAAU-VER-110-NAP
Supplier:  IBS Stock Items