ContactUsLinkComponent
Sucrose Standard 11.5% - 1.350149 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals place. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS115
Supplier:  REAGECON
Sucrose Standard 11.5% - 1.350149 ( 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals place. Extended shelf-life of 12 weeks 1 * 15 ml
Catalog # REGEBS115
Supplier:  REAGECON