Orders LinkContactUsLinkComponent
Flange clamps
Flange clamps
Ball and flat flanges KF/PF 50
Catalog # LENZ06248550
Supplier:   Lenz Laborglas
Flange clamps
Catalog # LENZ06248550
Supplier:  Lenz Laborglas
Flange clamps
Flange clamps
Ball and flat flanges KF/PF 50
Price
170,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Plastic, black.
  • Easy to assemble and disassemble
  • Tough construction, good temperature resistance
  • Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.
Specifications
  • For:
    Ball and flat flanges KF/PF 50
  • Seq:
    03