Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ENDOTOXIN-FREE (<0.005 EU/ML) 10XTUBES 1 * 1 KIT
Catalog # GENSM01072-10
Supplier:
Genscript
Expand image
ENDOTOXIN-FREE (<0.005 EU/ML) 10XTUBES 1 * 1 KIT
Catalog # GENSM01072-10
Supplier:
Genscript