Orders LinkContactUsLinkComponent
Adapters, delivery, straight
Adapters, delivery, straight
29/32 NS, 100 mm
Catalog # WITG1.900.029
Supplier:   witeg Labortechnik
Adapters, delivery, straight
Catalog # WITG1.900.029
Supplier:  witeg Labortechnik
Adapters, delivery, straight
Adapters, delivery, straight
29/32 NS, 100 mm
Price
22,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Socket size:
    29/32 NS
  • Tube length:
    100 mm