Orders LinkContactUsLinkComponent
Shelving systems, solid, Super Erecta®
Shelving systems, solid, Super Erecta®
Super Erecta® solid flat shelf, (width×length): 610×1524 mm, 2460FG, Galvanised steel, 14,0 kg
  139-0340
Shelving systems, solid, Super Erecta®
  139-0340
Shelving systems, solid, Super Erecta®
Shelving systems, solid, Super Erecta®
Super Erecta® solid flat shelf, (width×length): 610×1524 mm, 2460FG, Galvanised steel, 14,0 kg
323,00 €
1 * 1 items
Perfect for applications involving spillage. Provides barrier between floor and bottom shelf contents to deter contamination.
  • Shelf design features a 3,2 mm raised 'ship’s edge' on all four sides to contain spills
  • Louvered/embossed shelf allows air circulation
  • Available in several styles and materials for a variety of applications
  • Shelves are constructed with 18-gauge stainless steel (type 304) or galvanized materials
Standard stainless steel solid shelves (Type 304): with epoxy-coated cast corners address the majority of applications for solid, corrosion resistant shelves or work surfaces. Available in flat solid and louvered/embossed styles.
Galvanized shelves: with uncoated cast corners are ideal for applications requiring a solid shelving or work surface and minimum resistance to corrosion. Available in flat solid and louvered/embossed styles.
Premium Autoclave/Cart Washable Stainless Steel solid shelves (Type 304): feature an all-stainless construction to withstand high temperature, corrosive applications. Available in flat solid style.
Ordering information: Each individual shelf and post is sold separately. Accessories are also available.
  • Description:
    Super Erecta® solid flat shelf, (width×length): 610×1524 mm, 2460FG
  • Material:
    Galvanised steel
  • Seq:
    49
  • Weight:
    14,0 kg
Accessories
  •  
    Description
  • METO9990P
    Label holders, for 75×32 mm labels
    1 * 1 items - 8,30 €
  •  
    Description
    For
  • 139-0268
    Mobile post, SiteSelect™, chrome plated, 1590 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 50,10 €
  •  
    Description
    For
  • 139-0270
    Mobile post, SiteSelect™, chrome plated, 2454 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 69,60 €
  •  
    Description
    For
  • 139-0277
    Mobile post, SiteSelect™, stainless steel, 2454 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 items - 188,00 €
  •  
    Description
  • METO9990SL
    Slanted label holders, for 75×32 mm labels
    1 * 1 items - 8,30 €
  •  
    Description
    For
  • 139-0275
    Mobile post, SiteSelect™, stainless steel, 1590 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 items - 139,00 €
  •  
    Description
    For
  • 139-0255
    Stationary post, SiteSelect™, chrome plated, 2200 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 69,60 €
  •  
    Description
    For
  • 139-0251
    Stationary post, SiteSelect™, chrome plated, 875 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 42,60 €
  •  
    Description
  • METO2424BWI
    Decorator shelf inlays, black/white, 610×610 mm
    1 * 1 items - 71,30 €
  •  
    Description
  • METO9985
    Plastic split sleeves, black
    1 * 4 items - 15,00 €
  •  
    Description
  • METO9990CL5
    Clear label holders,1520 mm, for 1395×32 mm labels
    1 * 1 items - 42,00 €
  •  
    Description
    For
  • 139-0269
    Mobile post, SiteSelect™, chrome plated, 1880 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 54,70 €
  •  
    Description
  • METO9986S
    Aluminum split sleeves with stainless rings
    1 * 1 Each - 120,00 €
  •  
    Description
    For
  • 139-0276
    Mobile post, SiteSelect™, stainless steel, 1880 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 items - 152,00 €
  •  
    Description
    For
  • 139-0260
    Stationary post, SiteSelect™, stainless steel, 1590 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Stationary shelving, Super Erecta Pro™ shelving, Super Erecta Pro™ shelving
    1 * 1 items - 139,00 €
  •  
    Description
    For
  • METO63PK3
    Stationary post, SiteSelect™, metroseal 3, 1590 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
    1 * 1 items - 58,70 €
  •  
    Description
  • METOEP36C
    Enclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mm
    1 * 1 items - 133,00 €
  •  
    Description
    For
  • 139-0253
    Stationary post, SiteSelect™, chrome plated, 1590 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 50,10 €
  •  
    Description
    For
  • METO74PK3
    Stationary post, SiteSelect™, metroseal 3, 1895 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
    1 * 1 items - 63,90 €
  •  
    Description
  • METO2WS21C
    Post mounting shelf supports, width 530 mm, double, chrome plated
    1 * 1 items - 119,00 €
  •  
    Description
    For
  • 139-0254
    Stationary post, SiteSelect™, chrome plated, 1895 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 54,70 €
  •  
    Description
  • METO2WS14C
    Post mounting shelf supports, width 355 mm, double, chrome plated
    1 * 1 items - 113,00 €
  •  
    Description
  • METO1WS21C
    Post mounting shelf supports, width 530 mm, single, chrome plated
    1 * 1 items - 80,00 €
  •  
    Description
  • METO1WS14C
    Post mounting shelf supports, width 355 mm, single, chrome plated
    1 * 1 items - 77,10 €
  •  
    Description
  • METOH212C
    Storage baskets, chrome plated, 190×440×225 mm, 5 kg
    1 * 1 items - 106,00 €
  •  
    Description
  • METOH209C
    Storage baskets, chrome plated, 127×345×180 mm, 4 kg
    1 * 1 items - 75,90 €
  •  
    Description
  • METOEP57C
    Enclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mm
    1 * 1 items - 167,00 €
  •  
    Description
  • METO9084Z
    Additional tabs for rod, bag of 12
    1 * 1 Each - 19,00 €
  •  
    Description
    Material
  • METO1436CI-4
    Clear shelf inlays for Super Erecta® shelving system, 914×355 mm
    Plastic
    1 * 1 Each - 137,00 €
  •  
    Description
  • METOH118C
    Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point.
    1 * 1 items - 40,30 €
  •  
    Description
  • METO9986Z
    Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf)
    1 * 1 Each - 20,20 €
  •  
    Description
  • METOHHK6C
    Extension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandise
    1 * 1 items - 21,30 €
  •  
    Description
  • METO9985W
    Split sleeves, plastic, white
    1 * 1 Each - 15,00 €
  •  
    Description
  • METO9990CL
    Slanted label holders, for 75×32 mm labels
    1 * 1 items - 8,30 €
  •  
    Description
  • METO9990CL3
    Clear label holders, plastic, fits 914 mm shelfs, 725×32 mm
    1 * 1 items - 24,80 €
  •  
    Description
    Material
  • METO2136CI-4
    Clear shelf inlays for Super Erecta® shelving system, 914×530 mm
    Plastic
    1 * 1 Each - 148,00 €
  •  
    Description
    For
  • METO63UPS-SW
    Swaged post, stainless steel with aluminium cap, 1600 mm dimension
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
    1 * 1 items - 152,00 €
  •  
    Description
    For
  • METO96P
    Stationary post, SiteSelect™, chrome plated, 2454 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 79,40 €
  •  
    Description
    For
  • 139-0272
    Mobile post, SiteSelect™, stainless steel, 699 mm height
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 items - 98,40 €
  •  
    Description
    For
  • 139-0258
    Stationary post, SiteSelect™, stainless steel, 875 mm height
    Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 items - 112,00 €
  •  
    Description
    For
  • 139-0257
    Stationary post, SiteSelect™, stainless steel, 875 mm height
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 items - 98,40 €
  •  
    Description
    For
  • 139-0267
    Mobile post, SiteSelect™, chrome plated, 1385 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 47,80 €
  •  
    Description
    For
  • 139-0273
    Mobile post, SiteSelect™, stainless steel, 857 mm height
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 items - 112,00 €
  •  
    Description
    For
  • 139-0252
    Stationary post, SiteSelect™, chrome plated, 1385 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 items - 47,80 €
  •  
    Description
    For
  • 139-0262
    Stationary post, SiteSelect™, stainless steel, 2200 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 179,00 €
  •  
    Description
    For
  • 139-0263
    Mobile post, SiteSelect™, chrome plated, 191 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 29,40 €
  •  
    Description
    For
  • 139-0248
    Stationary post, SiteSelect™, chrome plated, 191 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 29,40 €
  •  
    Description
    For
  • 139-0274
    Mobile post, SiteSelect™, stainless steel, 1385 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 items - 122,00 €
  •  
    Description
    For
  • 139-0271
    Mobile post, SiteSelect™, stainless steel, 370 mm height
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    1 * 1 items - 74,20 €
  •  
    Description
    For
  • 139-0264
    Mobile post, SiteSelect™, chrome plated, 370 mm height
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    1 * 1 items - 32,80 €
  •  
    Description
    For
  • 139-0256
    Stationary post, SiteSelect™, stainless steel, 370 mm height
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    1 * 1 items - 74,20 €
  •  
    Description
    For
  • 139-0261
    Stationary post, SiteSelect™, stainless steel, 1895 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Stationary shelving, Super Erecta Pro™ shelving
    1 * 1 items - 152,00 €
  •  
    Description
  • METO9994Z
    Post clamps, joins units together for maximum strength, zinc-plated
    1 * 1 items - 14,40 €
  •  
    Description
    For
  • METO74UPK3
    Mobile post, SiteSelect™, metroseal 3, 1880 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
    1 * 1 items - 63,90 €
  •  
    Description
    For
  • 139-0265
    Mobile post, SiteSelect™, chrome plated, 699 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 39,70 €
  •  
    Description
    For
  • 139-0259
    Stationary post, SiteSelect™, stainless steel, 1385 mm height
    Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 items - 122,00 €
  •  
    Description
    For
  • 139-0250
    Stationary post, SiteSelect™, chrome plated, 699 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 items - 39,70 €
  •  
    Description
  • METO9986HZ
    Heavy duty aluminum split sleeves with zinc rings
    1 * 1 items - 52,90 €