Orders LinkContactUsLinkComponent
Cut resistant gloves, Sharpflex Nit
Cut resistant gloves, Sharpflex Nit
10, Green/black, 250 mm
Catalog # DALL2232524/10
Supplier:   HONEYWELL SAFETY
Cut resistant gloves, Sharpflex Nit
Catalog # DALL2232524/10
Supplier:  HONEYWELL SAFETY
Cut resistant gloves, Sharpflex Nit
Cut resistant gloves, Sharpflex Nit
10, Green/black, 250 mm
Price
Inquire For Price
Log in for availability
1 * 1 PAIR
Product Details & Documents
These gloves knitted from composite and para-aramid yarn features a black nitrile foam coating on the palm.
  • Knitting technique provides the glove with excellent fit and high dexterity
  • Para-aramid yarn protects against contact heat and cuts
  • Plated polyamide inside: Limit the contact of the skin with composite fiber
  • Nitrile coating provides good protection against oil and grease
  • Foam coating is more flexible than standard nitrile coating
Ideal for handling heavy parts with high cut risks in oily and greasy environments.
 : EN 388:2016 (3X42C), EN 407 (X1XXXX); Cat. II
Specifications
  • Glove size:
    10
  • Colour:
    Green/black
  • Length:
    250 mm
  • Seq:
    04
  • Cat. No.:
    DALL2232524/10
  • Liner material:
    Para-aramid
  • Disposable / reusable:
    Reusable
  • Model:
    Nit
  • Cuff style:
    Knit wrist
  • EN norm:
    EN 388:2016 (3X42C), EN 407 (X1XXXX)
  • Finish:
    Foam coating on palm and fingertips
  • Ambidextrous / hand-specific:
    Hand-specific
  • Gauge:
    13