Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FrameStar 480, 384well Plates, clear frame, white wells plus Q-PCR adhesive seals 1 * 50 ST
Catalog # FTIT0382
Supplier:
4titude Ltd.
Expand image
FrameStar 480, 384well Plates, clear frame, white wells plus Q-PCR adhesive seals 1 * 50 ST
Catalog # FTIT0382
Supplier:
4titude Ltd.