Order Entry
Export
Orders LinkContactUsLinkComponent
Reagents for comparators

Reagents for comparators

pH (phenol red), 250 tests, Phenol red, 1 KIT
Catalog # PALIAT130
Supplier:   Palintest
Reagents for comparators
Catalog # PALIAT130
Supplier:  Palintest
CAS Number:  
Reagents for comparators
Reagents for comparators

pH (phenol red), 250 tests, Phenol red, 1 KIT

CAS Number:  
Price
33,30 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Comparator tablet reagents are designed for use with the contour comparator, standard comparator and other visual test platforms.

Specifications

Specifications

  • Chemical method:
    Phenol red
  • Pk:
    1 KIT
  • Description:
    pH (phenol red), 250 tests

Specifications