Technical pH buffer solutions, HI 5000 series
3,00, ±0,01, 25 °C, 500 mL
Catalog # HANNHI5003
Supplier: Hanna
Jump to:
Product Details & Documents
- Colour coded labels
- ±0,01 pH
: Technical buffer solutions with certificate.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0,01
- pH value:3,00



