Funnels, powder
80 mm, 32 mm, 12 mm
Catalog # KAVA632413102080
Supplier: KAVALIER GLASS
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Ordering information: Ø = internal stem diameter.
Specifications
- Ø:12 mm
- Funnel O-Ø:80 mm
- Stem length:32 mm



