Ice Pan Lab with lid, Magic Touch 2, Mini, blue, 1.0L, expanded urethane, lightweight, chemical-resistant, snug lid, pour spout, fil l line indicator, raised top edge, easy transport, textured exterior, non-slip finish, 7.2 x 7.2 x5.3inches. 1 * 12 items
Catalog # BELAM168071191
Supplier: Bel-Art Products, a Part of SP