Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
HALOPERIDOL FOR SYSTEM SUITABILITY 1 * 25 mg
HALOPERIDOL FOR SYSTEM SUITABILITY 1 * 25 mg
Print
HALOPERIDOL FOR SYSTEM SUITABILITY 1 * 25 mg
Catalog # EDQMY0001517
Supplier:
EDQM
HALOPERIDOL FOR SYSTEM SUITABILITY 1 * 25 mg
Catalog # EDQMY0001517
Supplier:
EDQM
CAS Number:
Expand image
Return to Top
HALOPERIDOL FOR SYSTEM SUITABILITY 1 * 25 mg
Add to cart
CAS Number:
Price
270,00 €
Log in
for availability
Unit of Measure
1 * 25 mg
-
+
Add to cart
Return Policy
Product Details & Documents