Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Analytical column chiralpak® if 50 x 4.6 mm 5 µm 1 * 1 items
Catalog # DAIC86322
Supplier:
CHIRAL TECHNOLOGIES
Expand image
Analytical column chiralpak® if 50 x 4.6 mm 5 µm 1 * 1 items
Catalog # DAIC86322
Supplier:
CHIRAL TECHNOLOGIES