Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, printed label, sterile, Translucent, PP/PE
Catalog # 216-0987
Supplier: DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Catalog # 216-0987
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
PP translucent or PS clear, with PE screw cap.
- Manufactured from FDA approved virgin polystyrene or polypropylene
- PS offers good thermal and chemical resistance
- Supplied sterile - for in vitro use only
- All materials are non cytotoxic
Straight walled containers ideal for sample collection, storage and transport. Supplied with leakproof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certifications: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Packaging: Packed in inner bags of 50.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Type:Universal container, printed label, sterile
- Capacity:30 ml
- Colour:Translucent
- Material:PP/PE