Electrode solutions and reagents for ion selective electrodes
Chloride ionic strength adjustor (ISA) buffer powder pillows, 100
Catalog # HACH2318069
Supplier: Hach
Electrode solutions and reagents for ion selective electrodes
Catalog # HACH2318069
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:100
- Description:Chloride ionic strength adjustor (ISA) buffer powder pillows