Slide mailers
Cubic slide mailers with closure, Translucent, PP, 5
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Slide mailers made of polyethylene for archiving and staining of microscope slides.
- Secure closure and space for ²/₅ slides
- Break resistant rails for easy accommodation
- Transport of slides without contacting each other
Documents
Specifications
- No. of slides:5
- Colour:Translucent
- Description:Cubic slide mailers with closure
- Material:PP
- Seq:02